Ethyl oxalyl chloride ≥98%
Supplier: BeanTown Chemical
Synonyms:
Ethyl oxalyl monochloride, Oxalic Acid Monoethyl Ester Chloride, Ethyl Chlorooxoacetate, Ethyl Chloroglyoxylate, Chloroglyoxylic Acid Ethyl Ester, Ethyl chloroformylformate
139740-100G
139740-25G
139740-500G
BT139740-100GEA
83
USD
BT139740-100G
BT139740-25G
BT139740-500G
Ethyl oxalyl chloride ≥98%
Ethyl oxalyl chloride
CAS: 4755-77-5; EC No: 225-285-0; MDL No: MFCD00000706
UN No: UN2920; Haz Class: 8 (3); Packing Group: II
Liquid; Linear Formula: ClCOCOOCH2CH3; Molecular Formula: C4ClH5O3; MW: 136.53
Boiling Point: 135°; Flash point: 41°C (106°F)
Density (g/mL): 1.222; Refractive Index: 1.416
Moisture Sensitive
- Refractive index: 1.416
- Moisture Sensitive
- Liquid
Formula:
C₄H₅ClO₃ Boiling Pt: 135 °C Density: 1.222 g/mL Flash Pt: 41 °C (106 °F) |
MDL Number:
MFCD00000706 CAS Number: 4755-77-5 EINECS: 225-285-0 UN: 2920 ADR: 8,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...