Sodium fluoride ≥99.99% (trace metals basis)
Supplier: BeanTown Chemical
139730-100G
139730-25G
139730-5G
BT139730-100GEA
195.28
USD
BT139730-100G
BT139730-25G
BT139730-5G
Sodium fluoride ≥99.99% (trace metals basis)
Sodium fluoride
CAS: 7681-49-4; EC No: 231-667-8; MDL No: MFCD00003524; RTECS: WB0350000
UN No: UN1690; Haz Class: 6.1; Packing Group: III
Powder; Linear Formula: NaF; MW: 41.99
Melting Point: 993°; Boiling Point: 1704°
Density (g/mL): 2.78; Refractive Index: 1.325
Moisture Sensitive
- RTECS: WB0350000
- Refractive index: 1.325
- Moisture Sensitive
- Powder
Formula:
NaF MW: 41.99 g/mol Boiling Pt: 1704 °C Melting Pt: 993 °C Density: 2.78 g/mL |
MDL Number:
MFCD00003524 CAS Number: 7681-49-4 EINECS: 231-667-8 UN: 1690 ADR: 6.1,III |
Specification Test Results
Assay | 99.99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...