Titanium(IV) chloride ≥99.99% (trace metals basis)
Supplier: BeanTown Chemical
139660-100G
139660-25G
139660-5G
BT139660-100GEA
265
USD
BT139660-100G
BT139660-25G
BT139660-5G
Titanium(IV) chloride ≥99.99% (trace metals basis)
Titanium(IV) chloride
Formula:
Cl₄Ti Boiling Pt: 136…137 °C Melting Pt: –25 °C Density: 1.726 g/mL |
MDL Number:
MFCD00011267 CAS Number: 7550-45-0 EINECS: 231-441-9 UN: 1838 ADR: 8,I |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...