3,4-Difluorobenzonitrile ≥98%

Supplier: BeanTown Chemical

139625-100G 139625-25G 139625-5G
BT139625-100GEA 640 USD
BT139625-100G BT139625-25G BT139625-5G
3,4-Difluorobenzonitrile ≥98%
3,4-Difluorobenzonitrile

CAS: 64248-62-0; EC No: 264-751-8; MDL No: MFCD00011666 UN No: UN3439; Haz Class: 6.1; Packing Group: III Powder; Molecular Formula: C7H3F2N; MW: 139.10 Melting Point: 52-54°; Flash point: 69°C (156°F)


  • Powder

Formula: C₇H₃F₂N
MW: 139.1 g/mol
Melting Pt: 52…54 °C
Flash Pt: 69 °C (156 °F)
MDL Number: MFCD00011666
CAS Number: 64248-62-0
EINECS: 264-751-8
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR