Pentadecafluorocaprylic acid ≥95%
Supplier: BeanTown Chemical
Synonyms:
Pentadecafluorooctoic acid, Pentadecafluorooctanoic acid, PFOA, Perfluorocaprylic acid, Perfluorooctanoic acid
139610-25G
139610-5G
BT139610-25GEA
128.92
USD
BT139610-25G
BT139610-5G
Pentadecafluorocaprylic acid ≥95%
Pentadecafluorocaprylic acid
CAS: 335-67-1; EC No: 206-397-9; MDL No: MFCD00004174; RTECS: RH0781000
UN No: UN3261; Haz Class: 8; Packing Group: III
Flakes; Linear Formula: CF3(CF2)6COOH; Molecular Formula: C8HF15O2; MW: 414.07
Melting Point: 55-56°; Boiling Point: 189-192°
- RTECS: RH0781000
- Flakes
Formula:
C₈HF₁₅O₂ MW: 414.07 g/mol Boiling Pt: 189…192 °C Melting Pt: 55…56 °C |
MDL Number:
MFCD00004174 CAS Number: 335-67-1 EINECS: 206-397-9 UN: 3261 ADR: 8,III |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...