Ammonium peroxodisulphate (APS) ≥98.0% ACS
Supplier: BeanTown Chemical
Synonyms:
Peroxidisulphuric acid diammonium salt, Ammonium persulphate
139605-1KG
139605-250G
BT139605-1KGEA
110.52
USD
BT139605-1KG
BT139605-250G
Ammonium peroxodisulphate (APS) ≥98.0% ACS
Ammonium peroxodisulphate (APS)
CAS: 7727-54-0; EC No: 231-786-5; MDL No: MFCD00003390; RTECS: SE0350000
UN No: UN1444; Haz Class: 5.1; Packing Group: III
Lump; Linear Formula: (NH4)2S2O8; MW: 228.20
Moisture Sensitive
- RTECS: SE0350000
- Moisture Sensitive
- Lump
Formula:
(NH₄)₂S₂O₈ |
MDL Number:
MFCD00003390 CAS Number: 7727-54-0 EINECS: 231-786-5 UN: 1444 ADR: 5.1,III |
|
Specification Test Results
Assay | 98.0% min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...