Methacrylic acid ≥99% stabilized
Supplier: BeanTown Chemical
Synonyms:
2-Methylpropenoic acid
139535-2.5KG
139535-500G
BT139535-2.5KGEA
150.08
USD
BT139535-2.5KG
BT139535-500G
Methacrylic acid ≥99% stabilized
Methacrylic acid
CAS: 79-41-4; EC No: 201-204-4; MDL No: MFCD00002651; RTECS: OZ2975000
UN No: UN2531; Haz Class: 8; Packing Group: II
Liquid; Linear Formula: H2C=C(CH3)COOH; Molecular Formula: C4H6O2; MW: 86.09
Melting Point: 12-16°; Boiling Point: 163°; Flash point: 65°C (149°F)
Density (g/mL): 1.015; Refractive Index: 1.431
Moisture Sensitive
- RTECS: OZ2975000
- Refractive index: 1.431
- Moisture Sensitive
- Liquid
Formula:
C₄H₆O₂ MW: 86.09 g/mol Boiling Pt: 163 °C Melting Pt: 12…16 °C Density: 1.015 g/mL Flash Pt: 65 °C (149 °F) |
MDL Number:
MFCD00002651 CAS Number: 79-41-4 EINECS: 201-204-4 UN: 2531 ADR: 8,II |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...