2-Chlorothioxanthone ≥99%

Supplier: BeanTown Chemical

Synonyms: 2-Chlorothioxanthen-9-one, 2-Chloro-9H-thioxanthen-9-one

139515-100G 139515-25G 139515-5G
BT139515-100GEA 219 USD
BT139515-100G BT139515-25G BT139515-5G
2-Chlorothioxanthone ≥99%
2-Chlorothioxanthone

CAS: 86-39-5; EC No: 201-667-2; MDL No: MFCD00005067 UN No: UN1325; Haz Class: 4.1; Packing Group: III Powder; Molecular Formula: C13H7ClOS; MW: 246.71 Melting Point: 153-154°; Flash point: 91°C (196°F)


  • Powder

Formula: C₁₃H₇ClOS
MW: 246.72 g/mol
Melting Pt: 153…154 °C
Flash Pt: 91 °C (196 °F)
MDL Number: MFCD00005067
CAS Number: 86-39-5
EINECS: 201-667-2
ADR: 4.1,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR