Hexamethylenetetramine ≥99.0% ACS
Supplier: BeanTown Chemical
Synonyms:
Hexamethylenetetramine, Hexamine, Urotropine, Formin
139510-100G
139510-2KG
139510-500G
BT139510-100GEA
23.07
USD
BT139510-100G
BT139510-2KG
BT139510-500G
Hexamethylenetetramine ≥99.0% ACS
Hexamethylenetetramine
CAS: 100-97-0; EC No: 202-905-8; MDL No: MFCD00006895; RTECS: MN4725000
UN No: UN1328; Haz Class: 4.1; Packing Group: III
Solid; Molecular Formula: C6H12N4; MW: 140.19
Melting Point: 280° (sublimes); Flash point: 250°C (482°F)
Hygroscopic
- RTECS: MN4725000
- Hygroscopic
- Solid
Formula:
C₆H₁₂N₄ MW: 140.19 g/mol Melting Pt: 280 °C (subl.) Flash Pt: 250 °C (482 °F) |
MDL Number:
MFCD00006895 CAS Number: 100-97-0 EINECS: 202-905-8 UN: 1328 ADR: 4.1,III |
|
Specification Test Results
Assay | 99.0% min. |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...