Aminophylline, anhydrous ≥98%

Supplier: BeanTown Chemical

Synonyms: 3,7-Dihydro-1,3-dimethylpurine-2,6-dione, complex with 1,2-ethanediamine (2:1), 1,3-Dimethyl-3,7-dihydro-1H-purine-2,6-dione hemiethane-1,2-diamine salt

139505-100G 139505-25G 139505-500G
BT139505-100GEA 80 USD
BT139505-100G BT139505-25G BT139505-500G
Aminophylline, anhydrous ≥98%
Aminophylline

CAS: 317-34-0; EC No: 206-264-5; MDL No: MFCD00013221; RTECS: XH5600000 UN No: UN2811; Haz Class: 6.1; Packing Group: III Powder; Molecular Formula: C7H8N4O2·0.5C2H8N2; MW: 210.21


  • RTECS: XH5600000
  • Powder

Formula: C₇H₈N₄O₂
MW: 420.43 g/mol
MDL Number: MFCD00013221
CAS Number: 317-34-0
EINECS: 206-264-5
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR