Triisopropylchlorosilane ≥97%
Supplier: BeanTown Chemical
Synonyms:
Chlorotriisopropylsilane, Triisopropylsilyl chloride, Triisopropylchlorosilane
139425-10G
139425-250G
139425-50G
BT139425-10GEA
78.78
USD
BT139425-10G
BT139425-250G
BT139425-50G
Triisopropylchlorosilane ≥97%
Triisopropylchlorosilane
CAS: 13154-24-0; MDL No: MFCD00009656
UN No: UN2987; Haz Class: 8; Packing Group: II
Liquid; Linear Formula: [(CH3)2CH]3SiCl; Molecular Formula: C9H21ClSi; MW: 192.81
Boiling Point: 198°/739 mmHg; Flash point: 64°C (147°F)
Density (g/mL): 0.901; Refractive Index: 1.452
Moisture Sensitive
- Refractive index: 1.452
- Moisture Sensitive
- Liquid
Formula:
C₉H₂₁ClSi MW: 192.8 g/mol Boiling Pt: 198 °C/739 mmHg Density: 0.901 g/mL Flash Pt: 64 °C (147 °F) |
MDL Number:
MFCD00009656 CAS Number: 13154-24-0 UN: 2987 ADR: 8,II |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...