Molybdenum ≥99.9% (trace metals basis), powder APS 2-4 µm
Supplier: BeanTown Chemical
139395-100G
139395-25G
BT139395-100GEA
127.31
USD
BT139395-100G
BT139395-25G
Molybdenum ≥99.9% (trace metals basis), powder APS 2-4 µm
Molybdenum
CAS: 7439-98-7; EC No: 231-107-2; MDL No: MFCD00003465; RTECS: QA4680000
UN No: UN3089; Haz Class: 4.1; Packing Group: III
Powder; Molecular Formula: Mo; MW: 95.94
Melting Point: 2617°; Boiling Point: 4612°
Density (g/mL): 10.3
- RTECS: QA4680000
- Powder
Formula:
Mo Boiling Pt: 4612 °C Melting Pt: 2617 °C Density: 10.3 g/mL |
MDL Number:
MFCD00003465 CAS Number: 7439-98-7 EINECS: 231-107-2 UN: 3089 ADR: 4.1,III |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...