2,2,3,3,4,4,4-Heptafluorobutylamine ≥97%

Supplier: BeanTown Chemical

139390-1G 139390-5G
BT139390-1GEA 81 USD
BT139390-1G BT139390-5G
2,2,3,3,4,4,4-Heptafluorobutylamine ≥97%
2,2,3,3,4,4,4-Heptafluorobutylamine

CAS: 374-99-2; EC No: 206-780-0; MDL No: MFCD00014817; RTECS: EO5190000 UN No: UN2735; Haz Class: 8; Packing Group: III Liquid; Linear Formula: CF3CF2CF2CH2NH2; Molecular Formula: C4H4F7N; MW: 199.07 Density (g/mL): 1.456


  • RTECS: EO5190000
  • Liquid

Formula: C₄H₄F₇N
MW: 199.07 g/mol
Density: 1.456 g/mL
MDL Number: MFCD00014817
CAS Number: 374-99-2
EINECS: 206-780-0
ADR: 8,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR