Dibutylamine ≥98%
Supplier: BeanTown Chemical
Synonyms:
Di-n-butylamine, N-butyl-1-propanamine
139385-2.5L
139385-500ML
BT139385-2.5LEA
155.45
USD
BT139385-2.5L
BT139385-500ML
Dibutylamine ≥98%
Dibutylamine
CAS: 111-92-2; EC No: 203-921-8; MDL No: MFCD00009429; RTECS: HR7780000
UN No: UN2248; Haz Class: 8 (3); Packing Group: II
Liquid; Linear Formula: (CH3CH2CH2CH2)2NH; Molecular Formula: C8H19N; MW: 129.25
Melting Point: -62°; Boiling Point: 159°; Flash point: 40°C (104°F)
Density (g/mL): 0.767; Refractive Index: 1.417
- RTECS: HR7780000
- Refractive index: 1.417
- Liquid
Formula:
C₈H₁₉N Boiling Pt: 159 °C Melting Pt: –62 °C Density: 0.767 g/mL Flash Pt: 40 °C (104 °F) |
MDL Number:
MFCD00009429 CAS Number: 111-92-2 EINECS: 203-921-8 UN: 2248 ADR: 8,II |
|
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...