2,2-Difluoro-1,3-benzodioxole ≥97%

Supplier: BeanTown Chemical

Synonyms: 2,2-Difluorobenzo[d][1,3]dioxole

139380-25G 139380-5G
BT139380-25GEA 304 USD
BT139380-25G BT139380-5G
2,2-Difluoro-1,3-benzodioxole ≥97%
2,2-Difluoro-1,3-benzodioxole

CAS: 1583-59-1; EC No: 216-431-4; MDL No: MFCD00236217 UN No: UN1993; Haz Class: 3; Packing Group: III Liquid; Molecular Formula: C7H4F2O2 ; MW: 158.10 Flash point: 129°C (264°F) Density (g/mL): 1.303; Refractive Index: 1.444


  • Refractive index: 1.444
  • Liquid

Formula: C₇H₄F₂O₂
MW: 158.1 g/mol
Density: 1.303 g/mL
Flash Pt: 129 °C (264 °F)
MDL Number: MFCD00236217
CAS Number: 1583-59-1
EINECS: 216-431-4
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR