Tetrafluoroboric acid 50% (w/w) in water

Supplier: BeanTown Chemical

139335-4X500G 139335-500G
BT139335-4X500EA 155 USD
BT139335-4X500 BT139335-500G
Tetrafluoroboric acid 50% (w/w) in water
Tetrafluoroboric acid

CAS: 16872-11-0; EC No: 240-898-3; MDL No: MFCD00011345 UN No: UN1775; Haz Class: 8; Packing Group: II Liquid; Molecular Formula: HBF4; MW: 87.81 Boiling Point: 130° Density (g/mL): 1.41


  • Liquid

Formula: HBF₄
Boiling Pt: 130 °C
Density: 1.41 g/mL
MDL Number: MFCD00011345
CAS Number: 16872-11-0
EINECS: 240-898-3
UN: 1775
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR