Pyridinium dichromate ≥98%
Supplier: BeanTown Chemical
139270-100G
139270-25G
BT139270-100GEA
79.77
USD
BT139270-100G
BT139270-25G
Pyridinium dichromate ≥98%
Pyridinium dichromate
CAS: 20039-37-6; EC No: 243-478-8; MDL No: MFCD00013105
UN No: UN3087; Haz Class: 5.1 (6.1); Packing Group: II
Solid; Molecular Formula: C10H10N2·H2Cr2O7; MW: 376.20
Melting Point: 152-153°
Density (g/mL): 1.713
Hygroscopic
- Hygroscopic
- Solid
Formula:
C₁₀N₁₂Cr₂N₂O₇ Melting Pt: 152…153 °C Density: 1.713 g/mL |
MDL Number:
MFCD00013105 CAS Number: 20039-37-6 EINECS: 243-478-8 UN: 3087 ADR: 5.1,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...