Boron ≥99% (trace metals basis), powder, crystalline amorphous -325 mesh

Supplier: BeanTown Chemical

139265-10G 139265-50G
BT139265-10GEA 136.03 USD
BT139265-10G BT139265-50G
Boron ≥99% (trace metals basis), powder, crystalline amorphous -325 mesh
Boron

CAS: 7440-42-8; EC No: 231-151-2; MDL No: MFCD00134034; RTECS: ED7350000 UN No: UN3178; Haz Class: 4.1; Packing Group: III Powder; Molecular Formula: B; MW: 10.81 Melting Point: 2076°; Boiling Point: 3927° Density (g/mL): 2.34


  • RTECS: ED7350000
  • Powder

Formula: B
MW: 10.81 g/mol
Boiling Pt: 3927 °C
Melting Pt: 2076 °C
Density: 2.34 g/mL
MDL Number: MFCD00134034
CAS Number: 7440-42-8
EINECS: 231-151-2
UN: 3178
ADR: 4.1 III
Merck Index: 13,01333

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR