Acetylferrocene ≥97%
Supplier: BeanTown Chemical
139235-100G
139235-25G
139235-5G
BT139235-100GEA
353
USD
BT139235-100G
BT139235-25G
BT139235-5G
Acetylferrocene ≥97%
Acetylferrocene
CAS: 1271-55-2; EC No: 215-043-2; MDL No: MFCD00001432; RTECS: OB3700000
UN No: UN3467; Haz Class: 6.1; Packing Group: II
Crystalline; Molecular Formula: C12H12FeO; MW: 228.07
Melting Point: 81-83°
- RTECS: OB3700000
- Crystalline
Formula:
C₁₂H₁₂FeO Melting Pt: 81…83 °C |
MDL Number:
MFCD00001432 CAS Number: 1271-55-2 EINECS: 215-043-2 UN: 3467 ADR: 6.1,II |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...