Nicotinaldehyde ≥97%
Supplier: BeanTown Chemical
Synonyms:
Pyridine-3-carboxaldehyde, 3-Pyridinecarboxaldehyde, 3-Pyridinecarbaldehyde
139225-250G
139225-50G
BT139225-250GEA
257
USD
BT139225-250G
BT139225-50G
Nicotinaldehyde ≥97%
Nicotinaldehyde
CAS: 500-22-1; EC No: 207-900-4; MDL No: MFCD00006382; RTECS: QS2980000
UN No: UN2924; Haz Class: 3(8); Packing Group: III
Liquid; Molecular Formula: C6H5NO; MW: 107.11
Melting Point: 8°; Boiling Point: 78-81°/10 mmHg; Flash point: 35°C (95°F)
Density (g/mL): 1.141; Refractive Index: 1.549
Heat Sensitive
- RTECS: QS2980000
- Refractive index: 1.549
- Heat Sensitive
- Liquid
Formula:
C₆H₅NO MW: 107.11 g/mol Boiling Pt: 78…81 °C/10 mmHg Melting Pt: 8 °C Density: 1.141 g/mL Flash Pt: 35 °C (95 °F) |
MDL Number:
MFCD00006382 CAS Number: 500-22-1 EINECS: 207-900-4 ADR: 3(8),III |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...