Oxalacetic acid ≥97%

Supplier: BeanTown Chemical

Synonyms: Oxaloacetic acid, 2-Succinic acid, Oxalacetic acid

139205-100G 139205-25G 139205-5G
BT139205-100GEA 582 USD
BT139205-100G BT139205-25G BT139205-5G
Oxalacetic acid ≥97%
Oxalacetic acid

CAS: 328-42-7; EC No: 206-329-8; MDL No: MFCD00002592 UN No: UN3261; Haz Class: 8; Packing Group: II Powder; Linear Formula: HOOCCH2COCOOH; Molecular Formula: C4H4O5; MW: 132.07 Melting Point: 161°; Flash point: 88°C (190°F)


  • Powder

Formula: C₄H₄O₅
MW: 132.07 g/mol
Melting Pt: 161 °C
Flash Pt: 88 °C (190 °F)
MDL Number: MFCD00002592
CAS Number: 328-42-7
EINECS: 206-329-8
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR