1,10-Phenanthroline, anhydrous ≥99%
Supplier: BeanTown Chemical
Synonyms:
o-Phenanthroline
139190-100G
139190-25G
139190-5G
BT139190-100GEA
379.01
USD
BT139190-100G
BT139190-25G
BT139190-5G
1,10-Phenanthroline, anhydrous ≥99%
1,10-Phenanthroline
CAS: 66-71-7; EC No: 200-629-2; MDL No: MFCD00011678; RTECS: SF8300000
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Solid; Molecular Formula: C12H8N2; MW: 180.21
Melting Point: 114-117°
Hygroscopic
- RTECS: SF8300000
- Hygroscopic
- Solid
Formula:
C₁₂H₈N₂ MW: 180.21 g/mol Melting Pt: 114…117 °C |
MDL Number:
MFCD00011678 CAS Number: 66-71-7 EINECS: 200-629-2 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Assay | 99% |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...