Bismuth(III) iodide ≥99.999%

Supplier: BeanTown Chemical

Synonyms: Bismuth triiodide

139185-1G 139185-25G 139185-5G
BT139185-1GEA 19 USD
BT139185-1G BT139185-25G BT139185-5G
Bismuth(III) iodide ≥99.999%
Bismuth(III) iodide

CAS: 7787-64-6; EC No: 232-127-4; MDL No: MFCD00010894 UN No: UN3260; Haz Class: 8; Packing Group: III Powder/Lump; Molecular Formula: BiI3; MW: 589.69 Melting Point: 408° Hygroscopic


  • Hygroscopic
  • Powder/Lump

Formula: BiI₃
MW: 589.69 g/mol
Melting Pt: 408
MDL Number: MFCD00010894
CAS Number: 7787-64-6
EINECS: 232-127-4
UN: 3260
ADR: 8 III
Merck Index: 12,01308

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR