1,4-Naphthoquinone, (max. 5% H₂O) ≥97% (dry weight)
Supplier: BeanTown Chemical
Synonyms:
1,4-Naphthalenedione, 1,4-Dihydro-1,4-diketonaphthalene
139125-100G
139125-2.5KG
139125-500G
BT139125-100GEA
25.57
USD
BT139125-100G
BT139125-2.5KG
BT139125-500G
1,4-Naphthoquinone, (max. 5% H₂O) ≥97% (dry weight)
1,4-Naphthoquinone
CAS: 130-15-4; EC No: 204-977-6; MDL No: MFCD00001676; RTECS: QL7175000
UN No: UN2811; Haz Class: 6.1; Packing Group: I
Crystalline; Molecular Formula: C10H6O2; MW: 158.15
Melting Point: 119-122°
Density (g/mL): 1.42
- RTECS: QL7175000
- Crystalline
Formula:
C₁₀H₆O₂ MW: 158.16 g/mol Boiling Pt: 298 Melting Pt: 124 Density: 1.42 g/mL Flash Pt: 141 |
MDL Number:
MFCD00001676 CAS Number: 130-15-4 EINECS: 204-977-6 UN: 2811 ADR: 6.1 I Merck Index: 12,06482 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...