Ethylenediamine ≥99%

Supplier: BeanTown Chemical

Synonyms: 1,2-Ethanediamine, 1,2-Diaminoethane

139075-100ML 139075-2.5L 139075-500ML
BT139075-100MLEA 23.38 USD
BT139075-100ML BT139075-2.5L BT139075-500ML
Ethylenediamine ≥99%
Ethylenediamine

CAS: 107-15-3; EC No: 203-468-6; MDL No: MFCD00008204; RTECS: KH8575000 UN No: UN1604; Haz Class: 8 (3); Packing Group: II Liquid; Linear Formula: NH2CH2CH2NH2; Molecular Formula: C2H8N2; MW: 60.10 Melting Point: 8-10°; Boiling Point: 118°; Flash point: 38°C (100°F) Density (g/mL): 0.899; Refractive Index: 1.457 Air Sensitive


  • RTECS: KH8575000
  • Refractive index: 1.457
  • Air Sensitive
  • Liquid

Formula: C₂H₈N₂
MW: 60.1 g/mol
Boiling Pt: 117
Melting Pt: 8.5
Density: 0.899 g/mL
Flash Pt: 38 °C (100 °F)
MDL Number: MFCD00008204
CAS Number: 107-15-3
EINECS: 203-468-6
UN: 1604
ADR: 8 II
Merck Index: 13,03829
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0cbd90391008b8df01337e52ed5760383c063baa&vid=9a49702fe7150a0d3cd56afb586972447ecb6f85&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR