Hydriodic acid 55 - 58% ACS
Supplier: BeanTown Chemical
CAS: 10034-85-2; EC No: 233-109-9; MDL No: MFCD00011347; RTECS: MW3760000
UN No: UN1787; Haz Class: 8; Packing Group: II
Liquid; Molecular Formula: HI; MW: 127.91
Boiling Point: 127°
Density (g/mL): 1.701
Light Sensitive
- RTECS: MW3760000
- Light Sensitive
- Liquid
Formula:
HI Boiling Pt: 127 °C Density: 1.701 g/mL |
MDL Number:
MFCD00011347 CAS Number: 10034-85-2 EINECS: 233-109-9 UN: 1787 ADR: 8,II |
Specification Test Results
Assay | 55-58% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...