Lead(II) sulfate

Supplier: BeanTown Chemical

139050-100G 139050-2.5KG 139050-500G
BT139050-100GEA 46 USD
BT139050-100G BT139050-2.5KG BT139050-500G
Lead(II) sulfate
Lead(II) sulphate

CAS: 7446-14-2; EC No: 231-198-9; MDL No: MFCD00011166 UN No: UN2291; Haz Class: 6.1; Packing Group: III Powder/Lump; Linear Formula: PbSO4; MW: 303.25 Density (g/mL): 6.2


  • Powder/Lump

Formula: O₄PbS
Density: 6.2 g/mL
MDL Number: MFCD00011166
CAS Number: 7446-14-2
EINECS: 231-198-9
UN: 2291
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR