3-Aminobenzylamine ≥99%
Supplier: BeanTown Chemical
Synonyms:
alpha-Amino-m-toluidine, 3-(Aminomethyl)aniline
139045-25G
139045-5G
BT139045-25GEA
153
USD
BT139045-25G
BT139045-5G
3-Aminobenzylamine ≥99%
3-Aminobenzylamine
CAS: 4403-70-7; EC No: 412-230-2; MDL No: MFCD00078355
UN No: UN3259; Haz Class: 8; Packing Group: II
Solid; Molecular Formula: H2NC6H4CH2NH2; MW: 122.17
Melting Point: 41-45°; Flash point:
- Solid
Formula:
C₇H₁₀N₂ MW: 122.17 g/mol Melting Pt: 41…45 °C Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00078355 CAS Number: 4403-70-7 EINECS: 412-230-2 ADR: 8,II |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...