Magnesium perchlorate hexahydrate ≥99%
Supplier: BeanTown Chemical
139035-1KG
139035-250G
BT139035-1KGEA
234.4
USD
BT139035-1KG
BT139035-250G
Magnesium perchlorate hexahydrate ≥99%
Magnesium perchlorate hexahydrate
CAS: 13446-19-0; EC No: 233-108-3; MDL No: MFCD00149778
UN No: UN1475; Haz Class: 5.1; Packing Group: II
Crystalline; Linear Formula: Mg(ClO4)2·6H2O; MW: 331.30
Melting Point: 185-190°
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
Cl₂MgO₈·6H₂O Melting Pt: 185…190 °C |
MDL Number:
MFCD00149778 CAS Number: 13446-19-0 EINECS: 233-108-3 UN: 1475 ADR: 5.1,II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...