Rhodium(III) nitrate (10 - 15% Rh) (w/w) in water

Supplier: BeanTown Chemical

Synonyms: Rhodium(III) nitrate

138955-C2G 138955-C500MG
BT138955-C2GEA 1983 USD
BT138955-C2G BT138955-C500
Rhodium(III) nitrate (10 - 15% Rh) (w/w) in water
Rhodium(III) nitrate

CAS: 10139-58-9; EC No: 233-397-6; MDL No: MFCD00011202; RTECS: VI9316000 UN No: UN3093; Haz Class: 8 (5.1); Packing Group: II Liquid; Linear Formula: Rh(NO3)3; MW: 288.92 Density (g/mL): 1.41


  • RTECS: VI9316000
  • Liquid

Formula: N₃O₉Rh
Density: 1.41 g/mL
MDL Number: MFCD00011202
CAS Number: 10139-58-9
EINECS: 233-397-6
ADR: 8 (5.1),II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR