Calcium ≥99.5% (trace metals basis), shot ≤1 cm, redistilled

Supplier: BeanTown Chemical

138950-250G 138950-50G
BT138950-250GEA 143.37 USD
BT138950-250G BT138950-50G
Calcium ≥99.5% (trace metals basis), shot ≤1 cm, redistilled
Calcium

CAS: 7440-70-2; EC No: 213-179-5; MDL No: MFCD00085314; RTECS: EV8040000 UN No: UN1401; Haz Class: 4.3; Packing Group: II Solid; Molecular Formula: Ca; MW: 40.08 Melting Point: 850°; Boiling Point: 1484° Density (g/mL): 1.54


  • RTECS: EV8040000
  • Solid

Formula: Ca
Boiling Pt: 1484 °C
Melting Pt: 850 °C
Density: 1.54 g/mL
MDL Number: MFCD00085314
CAS Number: 7440-70-2
EINECS: 213-179-5
UN: 1401
ADR: 4.3,II

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR