Silver cyanide ≥99%

Supplier: BeanTown Chemical

Synonyms: Argentous cyanide

138940-100G 138940-25G
BT138940-100GEA 630 USD
BT138940-100G BT138940-25G
Silver cyanide ≥99%
Silver cyanide

CAS: 506-64-9; EC No: 208-048-6; MDL No: MFCD00003409; RTECS: VW3850000 UN No: UN1684; Haz Class: 6.1; Packing Group: II Powder; Linear Formula: AgCN; MW: 133.89 Melting Point: 320° (decomposes) Density (g/mL): 3.95; Refractive Index: 1.685 Light Sensitive


  • RTECS: VW3850000
  • Refractive index: 1.685
  • Light Sensitive
  • Powder

Formula: AgCN
MW: 133.89
Melting Pt: 320 °C (dec.)
Density: 3.943
MDL Number: MFCD00003409
CAS Number: 506-64-9
EINECS: 208-048-6
UN: 1684
ADR: 6.1 II
Merck Index: 12,08655

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR