Osmium(III) chloride trihydrate ≥99.99% (trace metals basis)

Supplier: BeanTown Chemical

138890-1G 138890-250MG 138890-5G
BT138890-1GEA 211 USD
BT138890-1G BT138890-250MG BT138890-5G
Osmium(III) chloride trihydrate ≥99.99% (trace metals basis)
Osmium(III) chloride trihydrate

CAS: 135296-80-9; EC No: 236-587-7; MDL No: MFCD00011148 UN No: UN3260; Haz Class: 8; Packing Group: III Crystalline; Linear Formula: OsCl3·3H2O; MW: 350.60


  • Crystalline

Formula: Cl₃Os·H₂O
MDL Number: MFCD00011148
CAS Number: 135296-80-9
EINECS: 236-587-7
UN: 3260
ADR: 8,III

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR