Lead(II) perchlorate trihydrate ≥97.0% ACS

Supplier: BeanTown Chemical

Synonyms: Lead diperchlorate trihydrate

138855-100G 138855-500G
BT138855-100GEA 48 USD
BT138855-100G BT138855-500G
Lead(II) perchlorate trihydrate ≥97.0% ACS
Lead(II) perchlorate trihydrate

CAS: 13453-62-8; EC No: 237-125-7; MDL No: MFCD00149817 UN No: UN1470; Haz Class: 5.1 (6.1); Packing Group: II Crystalline; Linear Formula: Pb(ClO4)2·3H2O; MW: 460.15 Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: Cl₂O₈Pb·3H₂O
MW: 460.15 g/mol
MDL Number: MFCD00149817
CAS Number: 13453-62-8
EINECS: 237-125-7
UN: 1470
ADR: 5.1,II

Order Now

Specification Test Results

Assay 97.0% min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR