Methyl malonyl chloride ≥97%

Supplier: BeanTown Chemical

Synonyms: Methyl 3-chloro-3-oxopropanoate

138815-1G 138815-25G 138815-5G
BT138815-1GEA 18 USD
BT138815-1G BT138815-25G BT138815-5G
Methyl malonyl chloride ≥97%
Methyl malonyl chloride

CAS: 37517-81-0; EC No: 253-540-6; MDL No: MFCD00013657 UN No: UN3265; Haz Class: 8; Packing Group: II Liquid; Linear Formula: ClCOCH2COOCH3; Molecular Formula: C4H5ClO3; MW: 136.53 Boiling Point: 57-59°/12 mmHg; Flash point: 80°C (176°F) Density (g/mL): 1.273; Refractive Index: 1.432 Light Sensitive, Moisture Sensitive


  • Refractive index: 1.432
  • Light Sensitive, Moisture Sensitive
  • Liquid

Formula: C₄H₅ClO₃
MW: 136.53 g/mol
Boiling Pt: 57…59 °C/12 mmHg
Density: 1.273 g/mL
Flash Pt: 80 °C (176 °F)
MDL Number: MFCD00013657
CAS Number: 37517-81-0
EINECS: 253-540-6
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR