2,2-Dimethyl-4-pentenoic acid ≥95%
Supplier: BeanTown Chemical
Synonyms:
2,2-Dimethylpent-4-enoic acid
138805-1G
138805-5G
BT138805-1GEA
53
USD
BT138805-1G
BT138805-5G
2,2-Dimethyl-4-pentenoic acid ≥95%
2,2-Dimethyl-4-pentenoic acid
CAS: 16386-93-9; MDL No: MFCD00012304
UN No: UN3265; Haz Class: 8; Packing Group: II
Liquid; Linear Formula: H2C=CHCH2C(CH3)2CO2H ; Molecular Formula: C7H12O2; MW: 128.17
Boiling Point: 104-108°/20 mmHg; Flash point: 88°C (190°F)
Density (g/mL): 0.933; Refractive Index: 1.431
- Refractive index: 1.431
- Liquid
Formula:
C₇H₁₂O₂ MW: 128.17 g/mol Boiling Pt: 104…108 °C/20 mmHg Density: 0.933 g/mL Flash Pt: 88 °C (190 °F) |
MDL Number:
MFCD00012304 CAS Number: 16386-93-9 ADR: 8,II |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...