p-Toluenesulfonylsemicarbazide

Supplier: BeanTown Chemical

138795-100G 138795-25G
BT138795-100GEA 256 USD
BT138795-100G BT138795-25G
p-Toluenesulfonylsemicarbazide
p-Toluenesulfonylsemicarbazide

CAS: 10396-10-8; EC No: 233-857-6; MDL No: MFCD00072243 UN No: UN2811; Haz Class: 6.1; Packing Group: III Powder; Linear Formula: CH3C6H4SO2NHNHCONH2; Molecular Formula: C8H11N3O3S; MW: 229.26 Density (g/mL): 1.381


  • Powder

Formula: C8H11N3O3S
MW: 229.26 g/mol
Density: 1.381 g/mL
MDL Number: MFCD00072243
CAS Number: 10396-10-8
EINECS: 233-857-6
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR