4-Methoxybenzonitrile ≥99%

Supplier: BeanTown Chemical

Synonyms: 4-Cyanoanisol, Anisonitrile, p-Cyanoanisol, p-Methoxybenzonitrile

138770-100G 138770-25G 138770-5G
BT138770-100GEA 273 USD
BT138770-100G BT138770-25G BT138770-5G
4-Methoxybenzonitrile ≥99%
4-Methoxybenzonitrile

CAS: 874-90-8; EC No: 212-871-6; MDL No: MFCD00001818 UN No: UN3439; Haz Class: 6.1; Packing Group: III Solid; Molecular Formula: C8H7NO; MW: 133.15 Melting Point: 57-59°; Boiling Point: 256-257° Hygroscopic


  • Hygroscopic
  • Solid

Formula: C₈H₇NO
MW: 133.15 g/mol
Boiling Pt: 256…257 °C
Melting Pt: 57…59 °C
MDL Number: MFCD00001818
CAS Number: 874-90-8
EINECS: 212-871-6
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR