Morpholine ≥99%

Supplier: BeanTown Chemical

138755-100G 138755-2.5KG 138755-500G
BT138755-100GEA 29.78 USD
BT138755-100G BT138755-2.5KG BT138755-500G
Morpholine ≥99%
Morpholine

CAS: 110-91-8; EC No: 203-815-1; MDL No: MFCD00005972; RTECS: QD6475000 UN No: UN2054; Haz Class: 8 (3); Packing Group: I Liquid; Molecular Formula: C4H9NO; MW: 87.12 Melting Point: -7 - -5°; Boiling Point: 129°; Flash point: 31°C (88°F) Density (g/mL): 0.996; Refractive Index: 1.454 Hygroscopic


  • RTECS: QD6475000
  • Refractive index: 1.454
  • Hygroscopic
  • Liquid

Formula: C₄H₉NO
MW: 87.12 g/mol
Boiling Pt: 129 °C
Melting Pt: –7 – –5 °C
Density: 0.996 g/mL
Flash Pt: 31 °C (88 °F)
MDL Number: MFCD00005972
CAS Number: 110-91-8
EINECS: 203-815-1
UN: 2054
ADR: 8,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9fb9ce2adfd770be2ae8d5f63ea6220bef0f781&vid=6b2bfa885631c3f4e0681813373d468dcc171b83&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR