Iron ≥99.9% (trace metals basis), nanopowder APS 10-30 nm
Supplier: BeanTown Chemical
CAS: 7439-89-6; EC No: 231-096-4; MDL No: MFCD00010999; RTECS: NO4565500
UN No: UN3089 ; Haz Class: 4.1; Packing Group: II
Powder, FeO layer 0.5-1.5nm ; Molecular Formula: Fe; MW: 55.85
Melting Point: 1535°; Boiling Point: 2750°
Density (g/mL): 7.86
- RTECS: NO4565500
- Powder, FeO layer 0.5-1.5nm
Formula:
Fe Boiling Pt: 2750 °C Melting Pt: 1535 °C Density: 7.86 g/mL |
MDL Number:
MFCD00010999 CAS Number: 7439-89-6 EINECS: 231-096-4 UN: 3089 ADR: 4.1,II |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...