Iridium(IV) oxide ≥99% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Iridium dioxide, Iridium(IV) oxide

138700-1G 138700-5G
BT138700-1GEA 370.18 USD
BT138700-1G BT138700-5G
Iridium(IV) oxide ≥99% (trace metals basis)
Iridium(IV) oxide

CAS: 12030-49-8; EC No: 234-743-9; MDL No: MFCD00011065 UN No: UN1479; Haz Class: 5.1; Packing Group: II Powder; Molecular Formula: IrO2; MW: 224.21


  • Powder

Formula: IrO₂
MW: 224.22 g/mol
MDL Number: MFCD00011065
CAS Number: 12030-49-8
EINECS: 234-743-9
UN: 1479
ADR: 5.1,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR