2,6-Difluoropyridine ≥98%
Supplier: BeanTown Chemical
138695-100G
138695-25G
138695-5G
BT138695-100GEA
433
USD
BT138695-100G
BT138695-25G
BT138695-5G
2,6-Difluoropyridine ≥98%
2,6-Difluoropyridine
CAS: 1513-65-1; EC No: 216-152-8; MDL No: MFCD00006227
UN No: UN1993; Haz Class: 3; Packing Group: III
Liquid; Molecular Formula: C5H3F2N; MW: 115.08
Boiling Point: 125°/743 mmHg; Flash point: 33°C (91°F)
Density (g/mL): 1.268; Refractive Index: 1.437
- Refractive index: 1.437
- Liquid
Formula:
C₅H₃F₂N MW: 115.08 g/mol Boiling Pt: 125 °C/743 mmHg Density: 1.268 g/mL Flash Pt: 33 °C (91 °F) |
MDL Number:
MFCD00006227 CAS Number: 1513-65-1 EINECS: 216-152-8 UN: 1993 ADR: 3,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...