α,α'-Dibromo-p-xylene ≥97%
Supplier: BeanTown Chemical
Synonyms:
p-Xylylene dibromide, 1,4-Bis(bromomethyl)benzene
138630-10G
138630-50G
BT138630-10GEA
44
USD
BT138630-10G
BT138630-50G
α,α'-Dibromo-p-xylene ≥97%
α,α'-Dibromo-p-xylene
CAS: 623-24-5; EC No: 210-781-1; MDL No: MFCD00000182
UN No: UN3261; Haz Class: 8; Packing Group: II
Crystalline; Linear Formula: C6H4(CH2Br)2; Molecular Formula: C8H8Br2; MW: 263.96
Melting Point: 143-145°; Boiling Point: 245°
Density (g/mL): 2.012
- Crystalline
Formula:
C₈H₈Br₂ Boiling Pt: 245 °C Melting Pt: 143…145 °C Density: 2.012 g/mL |
MDL Number:
MFCD00000182 CAS Number: 623-24-5 EINECS: 210-781-1 UN: 3261 ADR: 8,II |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...