Cadmium (II) sulfate 8/3-hydrate 98.0-102.0% ACS
Supplier: BeanTown Chemical
138610-1KG
138610-250G
138610-25G
BT138610-1KGEA
347.94
USD
BT138610-1KG
BT138610-250G
BT138610-25G
Cadmium (II) sulfate 8/3-hydrate 98.0-102.0% ACS
Cadmium(II) sulphate 8/3-hydrate
CAS: 7790-84-3; EC No: 233-331-6; MDL No: MFCD00151655; RTECS: EV2850000
UN No: UN2570; Haz Class: 6.1; Packing Group: III
Crystalline; Molecular Formula: 3CdSO4·8H2O ; MW: 769.71
- RTECS: EV2850000
- Crystalline
Formula:
3CdSO₄·8H₂O |
MDL Number:
MFCD02253000 CAS Number: 7790-84-3 EINECS: 233-331-6 UN: 2570 ADR: 6.1,III |
Specification Test Results
Assay | 98.0-102.0% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...