4,4-Dimethyl-3-thiosemicarbazide ≥95%

Supplier: BeanTown Chemical

138575-1G 138575-5G
BT138575-1GEA 51 USD
BT138575-1G BT138575-5G
4,4-Dimethyl-3-thiosemicarbazide ≥95%
4,4-Dimethyl-3-thiosemicarbazide

CAS: 6926-58-5; MDL No: MFCD00041308 UN No: UN2811; Haz Class: 6.1; Packing Group: III Powder; Molecular Formula: C3H9N3S; MW: 119.19 Melting Point: 153° (decomposes)


  • Powder

Formula: C₃H₉N₃S
Melting Pt: 153 °C (dec.)
MDL Number: MFCD00041308
CAS Number: 6926-58-5
ADR: 6.1,III

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR