2-Pentanone ≥99%

Supplier: BeanTown Chemical

138500-100G 138500-25G 138500-500G
BT138500-100GEA 29.94 USD
BT138500-100G BT138500-25G BT138500-500G
2-Pentanone ≥99%
2-Pentanone

CAS: 107-87-9; EC No: 203-528-1; MDL No: MFCD00009400; RTECS: SA7875000 UN No: UN1249; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH3COCH2CH2CH3; Molecular Formula: C5H10O; MW: 86.13 Melting Point: -78°; Boiling Point: 101-105°; Flash point: 7°C (45°F) Density (g/mL): 0.809; Refractive Index: 1.390


  • RTECS: SA7875000
  • Refractive index: 1.390
  • Liquid

Formula: C₅H₁₀O
Boiling Pt: 101…105 °C
Melting Pt: –78 °C
Density: 0.809 g/mL
Flash Pt: 7 °C (45 °F)
MDL Number: MFCD00009400
CAS Number: 107-87-9
EINECS: 203-528-1
UN: 1249
ADR: 3,II

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR