p-Xylylenebis(triphenylphosphonium chloride) ≥97%
Supplier: BeanTown Chemical
138470-25G
138470-5G
BT138470-25GEA
216
USD
BT138470-25G
BT138470-5G
p-Xylylenebis(triphenylphosphonium chloride) ≥97%
p-Xylylenebis(triphenylphosphonium chloride)
CAS: 1519-47-7; EC No: 216-184-2; MDL No: MFCD00031710; RTECS: TA3580000
UN No: UN3464; Haz Class: 6.1; Packing Group: III
Solid; Molecular Formula: C44H38Cl2P2; MW: 699.64
Melting Point:
- RTECS: TA3580000
- Hygroscopic
- Solid
Formula:
C₄₄H₃₈Cl₂P₂ MW: 699.66 g/mol Melting Pt: >300 °C |
MDL Number:
MFCD00031710 CAS Number: 1519-47-7 EINECS: 216-184-2 UN: 3464 ADR: 6.1 III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...