3-Amino-4-methylpyridine ≥98%
Supplier: BeanTown Chemical
138460-25G
138460-5G
BT138460-25GEA
255
USD
BT138460-25G
BT138460-5G
3-Amino-4-methylpyridine ≥98%
3-Amino-4-methylpyridine
CAS: 3430-27-1; MDL No: MFCD00128871
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Powder; Molecular Formula: C6H8N2 ; MW: 108.14
Melting Point: 102-107°
Hygroscopic
- Hygroscopic
- Powder
Formula:
C₆H₈N₂ MW: 108.14 g/mol Melting Pt: 102…107 °C |
MDL Number:
MFCD00128871 CAS Number: 3430-27-1 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...