Sodium ethoxide ≥96%

Supplier: BeanTown Chemical

Synonyms: Sodium ethylate, Sodium ethanolate

138410-100G 138410-2KG 138410-500G
BT138410-100GEA 58.29 USD
BT138410-100G BT138410-2KG BT138410-500G
Sodium ethoxide ≥96%
Sodium ethoxide

CAS: 141-52-6; EC No: 205-487-5; MDL No: MFCD00012417 UN No: UN3206; Haz Class: 4.2 (8); Packing Group: II Powder; Linear Formula: CH3CH2ONa; MW: 68.05 Flash point: 30°C (86°F) Moisture Sensitive


  • Moisture Sensitive
  • Powder

Formula: C₂H₅NaO
MW: 68.05 g/mol
Melting Pt: 260
Flash Pt: 30 °C (86 °F)
MDL Number: MFCD00012417
CAS Number: 141-52-6
EINECS: 205-487-5
UN: 3206
ADR: 4.2 II

Order Now

Specification Test Results

Assay 96%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR