Iron(III)-2-ethylhexanoate 50% (w/w) in mineral spirits
Supplier: BeanTown Chemical
138390-1KG
138390-250G
138390-50G
BT138390-1KGEA
188
USD
BT138390-1KG
BT138390-250G
BT138390-50G
Iron(III)-2-ethylhexanoate 50% (w/w) in mineral spirits
Iron(III)-2-ethylhexanoate
CAS: 7321-53-1; EC No: 230-794-6; MDL No: MFCD00014006
UN No: UN1268; Haz Class: 3; Packing Group: III
Liquid; Molecular Formula: C24H45FeO6 ; MW: 485.47
Density (g/mL): 0.91
- Liquid
Formula:
C₂₄H₄₅FeO₆ Density: 0.91 g/mL |
MDL Number:
MFCD00014006 CAS Number: 7321-53-1 EINECS: 230-794-6 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...